Product Details

SNP ID
rs3767214
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:70149458 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
ACTAGAGAAGCAATAGCCAAGAGTA[C/T]AGATGGACCAAATGACCCCTTAATA
Phenotype
MIM: 614453
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC40 PubMed Links

Gene Details

Gene
LRRC40
Gene Name
leucine rich repeat containing 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017768.4 Intron NP_060238.3
XM_005271013.1 Intron XP_005271070.1
XM_011541763.1 Intron XP_011540065.1
XM_017001695.1 Intron XP_016857184.1
XM_017001696.1 Intron XP_016857185.1
Gene
LRRC7
Gene Name
leucine rich repeat containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020794.2 Intron NP_065845.1
XM_017001885.1 Intron XP_016857374.1
XM_017001886.1 Intron XP_016857375.1
XM_017001887.1 Intron XP_016857376.1
XM_017001888.1 Intron XP_016857377.1
XM_017001889.1 Intron XP_016857378.1
XM_017001890.1 Intron XP_016857379.1
XM_017001891.1 Intron XP_016857380.1
XM_017001892.1 Intron XP_016857381.1
XM_017001893.1 Intron XP_016857382.1
XM_017001894.1 Intron XP_016857383.1
XM_017001895.1 Intron XP_016857384.1

View Full Product Details