Product Details
- SNP ID
-
rs3771285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:47810771 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAATTCCTTTGAAATATTTCTAGA[A/G]CCTTTGTATCCTCTTTGTTTTTAGT
- Phenotype
-
MIM: 607871
MIM: 600678
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114518335] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO11
- Gene Name
- F-box protein 11
- Gene
- MSH6
- Gene Name
- mutS homolog 6
There are no transcripts associated with this gene.
View Full Product Details