Product Details

SNP ID
rs3786992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:802209 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGAGCAGGTGCCGGAGTCTCCC[G/A]AGGCTCTTGCCGTCCGCGTCGGGAG
Phenotype
MIM: 600693
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MIR4745 PubMed Links

Gene Details

Gene
MIR4745
Gene Name
microRNA 4745
There are no transcripts associated with this gene.

Gene
PTBP1
Gene Name
polypyrimidine tract binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002819.4 Intron NP_002810.1
NM_031990.3 Intron NP_114367.1
NM_031991.3 Intron NP_114368.1
XM_005259597.2 Intron XP_005259654.1
XM_005259598.2 Intron XP_005259655.1

View Full Product Details