Product Details

SNP ID
rs3787052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35711153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATCAATGAGCAAACCAGCGGGG[A/G]AGCTGAGTCACAAGCACATGATGTG
Phenotype
MIM: 606834 MIM: 605859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KMT2B PubMed Links

Gene Details

Gene
KMT2B
Gene Name
lysine methyltransferase 2B
There are no transcripts associated with this gene.

Gene
ZBTB32
Gene Name
zinc finger and BTB domain containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316902.1 Intron NP_001303831.1
NM_001316903.1 Intron NP_001303832.1
NM_014383.2 Intron NP_055198.1
XM_011526718.2 Intron XP_011525020.1
XM_017026588.1 Intron XP_016882077.1
XM_017026589.1 Intron XP_016882078.1
XM_017026590.1 Intron XP_016882079.1
XM_017026591.1 Intron XP_016882080.1
XM_017026592.1 Intron XP_016882081.1

View Full Product Details