Product Details

SNP ID
rs3797863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:162072449 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAATCAGAGTAGTTTGGAGATTG[C/T]TTGACCACTGTGGGTGTCGGCACAA
Phenotype
MIM: 137164
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRG2 PubMed Links

Gene Details

Gene
GABRG2
Gene Name
gamma-aminobutyric acid type A receptor gamma2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000816.3 Intron NP_000807.2
NM_198903.2 Intron NP_944493.2
NM_198904.2 Intron NP_944494.1

View Full Product Details