Product Details

SNP ID
rs3799990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105277685 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAAACTCCACTTCTGGAGTTGC[C/T]CCTCAATATGAAGAAACCGGAAGTA
Phenotype
MIM: 600400
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PREP PubMed Links

Gene Details

Gene
PREP
Gene Name
prolyl endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002726.4 2825 UTR 3 NP_002717.3
XM_005267044.2 2825 Intron XP_005267101.1
XM_011535925.2 2825 Intron XP_011534227.1

View Full Product Details