Product Details

SNP ID
rs3802776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95166683 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATCCAAGTAGCAAGGCCATGTT[C/G]CAAGTGGACCTGACAGGGTTCTTTA
Phenotype
MIM: 607768
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929295 PubMed Links

Gene Details

Gene
LOC101929295
Gene Name
uncharacterized LOC101929295
There are no transcripts associated with this gene.

Gene
SESN3
Gene Name
sestrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271594.1 8377 UTR 3 NP_001258523.1
NM_144665.3 8377 UTR 3 NP_653266.2
XM_011542614.1 8377 UTR 3 XP_011540916.1

View Full Product Details