Product Details

SNP ID
rs3814563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43646539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCCCCACCTAACCACCTACTA[C/T]TTTCTTCCCTCCAAGCCCCAGAGGA
Phenotype
MIM: 194539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF32 PubMed Links

Gene Details

Gene
ZNF32
Gene Name
zinc finger protein 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005368.2 2812 Intron NP_001005368.1
NM_001324164.1 2812 Intron NP_001311093.1
NM_001324165.1 2812 Intron NP_001311094.1
NM_001324166.1 2812 Intron NP_001311095.1
NM_001324167.1 2812 Intron NP_001311096.1
NM_001324168.1 2812 Intron NP_001311097.1
NM_006973.2 2812 Intron NP_008904.1
XM_017016616.1 2812 UTR 5 XP_016872105.1
Gene
ZNF32-AS1
Gene Name
ZNF32 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF32-AS2
Gene Name
ZNF32 antisense RNA 2
There are no transcripts associated with this gene.

Gene
ZNF32-AS3
Gene Name
ZNF32 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details