Product Details

SNP ID
rs3827804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:77462215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCAAAGGTCGCTTAGGCGACCC[C/T]ACTGCAAACGGCAAGTCAACTTGTG
Phenotype
MIM: 182131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR1B PubMed Links

Gene Details

Gene
HTR1B
Gene Name
5-hydroxytryptamine receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000863.2 1808 UTR 3 NP_000854.1

View Full Product Details