Product Details
- SNP ID
-
rs3207249
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:22447183 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCCCGGCGTTGCCGGTACCATCG[A/G]GGATGCAGGTTCCCCCAGCGTTCCT
- Phenotype
-
MIM: 614803
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DMRTA1
PubMed Links
Gene Details
- Gene
- DMRTA1
- Gene Name
- DMRT like family A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022160.2 |
343 |
Missense Mutation |
AGG,GGG |
R40G |
NP_071443.2 |
View Full Product Details