Product Details

SNP ID
rs3209642
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:53973108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTAGCGAAAATGAAGAGGAGA[C/T]AGAAGAGGAGCTACAAGGTGATGAT
Phenotype
MIM: 601425
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TCEA1 PubMed Links
Additional Information
For this assay, SNP(s) [rs148045853] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TCEA1
Gene Name
transcription elongation factor A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006756.3 Intron NP_006747.1
NM_201437.2 Intron NP_958845.1
XM_006716467.1 Intron XP_006716530.1
XM_011517578.1 Intron XP_011515880.1

View Full Product Details