Product Details

SNP ID
rs3732303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:75047000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACAGTACAAATCATGGTGTTTCT[C/T]ACATGTGGGAGTTGATGACATTTTT
Phenotype
MIM: 162323
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TACR1 PubMed Links

Gene Details

Gene
TACR1
Gene Name
tachykinin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001058.3 4241 UTR 3 NP_001049.1
NM_015727.2 4241 Intron NP_056542.1

View Full Product Details