Product Details

SNP ID
rs3738582
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:156153439 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGCCTCTTAAACTTTCTGGTTGGAC[C/G]GAGCAGAGGAGGGAGAGAGAGGTGT
Phenotype
MIM: 607292
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SEMA4A PubMed Links

Gene Details

Gene
SEMA4A
Gene Name
semaphorin 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193300.1 Intron NP_001180229.1
NM_001193301.1 Intron NP_001180230.1
NM_001193302.1 Intron NP_001180231.1
NM_022367.3 Intron NP_071762.2
XM_011509871.2 Intron XP_011508173.1
XM_011509872.2 Intron XP_011508174.1
XM_011509873.2 Intron XP_011508175.1
XM_011509874.2 Intron XP_011508176.1
XM_011509875.2 Intron XP_011508177.1
XM_011509876.2 Intron XP_011508178.1
XM_011509878.2 Intron XP_011508180.1
XM_011509879.2 Intron XP_011508181.1
XM_017002056.1 Intron XP_016857545.1
XM_017002057.1 Intron XP_016857546.1

View Full Product Details