Product Details

SNP ID
rs3741857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8032955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTGCCCAGGCCAGCTCAGGGA[C/G]AGCCGGGAGTACGAAAGCCCCACCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FOXJ2 PubMed Links

Gene Details

Gene
FOXJ2
Gene Name
forkhead box J2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018416.2 266 UTR 5 NP_060886.1
XM_011520760.2 266 UTR 5 XP_011519062.1
XM_011520761.2 266 UTR 5 XP_011519063.1
XM_017019640.1 266 Intron XP_016875129.1

View Full Product Details