Product Details
- SNP ID
-
rs3742786
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:74906308 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGACTTGCTCTAAGGGGCAGACCC[A/G]TGCATTCCTTCCTCCCCCCATTACT
- Phenotype
-
MIM: 126063
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DLST
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145681193] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DLST
- Gene Name
- dihydrolipoamide S-succinyltransferase
There are no transcripts associated with this gene.
- Gene
- RPS6KL1
- Gene Name
- ribosomal protein S6 kinase like 1
View Full Product Details