Product Details

SNP ID
rs3761054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17723989 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTTGAGTTTCTCTTTCTTGCCT[G/T]GTCCTGACCTGCAGCTCTGCCGTGC
Phenotype
MIM: 607573
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP1S PubMed Links

Gene Details

Gene
MAP1S
Gene Name
microtubule associated protein 1S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308363.1 Intron NP_001295292.1
NM_018174.5 Intron NP_060644.4
XM_017026930.1 Intron XP_016882419.1
XM_017026931.1 Intron XP_016882420.1

View Full Product Details