Product Details

SNP ID
rs3776140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:32125638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTATAGTTAAAATTATAACAAG[C/T]GTAATAATACAATAGATTTACATGG
Phenotype
MIM: 612207
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLPH3 PubMed Links

Gene Details

Gene
GOLPH3
Gene Name
golgi phosphoprotein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022130.3 2025 UTR 3 NP_071413.1
XM_017009686.1 2025 UTR 3 XP_016865175.1
XM_017009687.1 2025 UTR 3 XP_016865176.1

View Full Product Details