Product Details

SNP ID
rs3776423
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43487772 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGATCCATCTTAAATTACAATC[G/T]TAGGTATTTGCTGACTCAATAAAGG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
C5orf34 PubMed Links
Additional Information
For this assay, SNP(s) [rs77026722] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C5orf34
Gene Name
chromosome 5 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198566.3 3208 Intron NP_940968.1
XM_011514038.2 3208 UTR 3 XP_011512340.1
XM_017009445.1 3208 Intron XP_016864934.1
XM_017009446.1 3208 Intron XP_016864935.1
Gene
TMEM267
Gene Name
transmembrane protein 267
There are no transcripts associated with this gene.

View Full Product Details