Product Details

SNP ID
rs3806612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:97766169 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAGATGCTTAAAGGGGGGAAAAT[A/G]TGAGAAGTTGTCGTTTTTAAAACAT
Phenotype
MIM: 608845 MIM: 600066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL6 PubMed Links

Gene Details

Gene
ARL6
Gene Name
ADP ribosylation factor like GTPase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006713783.3 Intron XP_006713846.1
XM_017007311.1 Intron XP_016862800.1
XM_017007312.1 Intron XP_016862801.1
XM_017007313.1 Intron XP_016862802.1
Gene
EPHA6
Gene Name
EPH receptor A6
There are no transcripts associated with this gene.

View Full Product Details