Product Details
- SNP ID
-
rs3810908
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:113165995 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTCACCTTGACCTCACCCTGTGG[C/T]TCGCCTAGACCCAGGGTTGGCTGTG
- Phenotype
-
MIM: 603088
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FKBP15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs540268353] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FKBP15
- Gene Name
- FK506 binding protein 15
- Gene
- SLC31A2
- Gene Name
- solute carrier family 31 member 2
There are no transcripts associated with this gene.
View Full Product Details