Product Details

SNP ID
rs3826429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68869325 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAGTCAGTTTCTTAATATACAG[C/G]CTAAGACAGTAAAATAATATGCCAG
Phenotype
MIM: 612505
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABCA8 PubMed Links

Gene Details

Gene
ABCA8
Gene Name
ATP binding cassette subfamily A member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288985.1 Intron NP_001275914.1
NM_001288986.1 Intron NP_001275915.1
NM_007168.3 Intron NP_009099.1
XM_005256938.2 Intron XP_005256995.1
XM_005256940.4 Intron XP_005256997.1
XM_011524191.2 Intron XP_011522493.1
XM_011524192.2 Intron XP_011522494.1

View Full Product Details