Product Details

SNP ID
hCV27528994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48565160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGTACTCCTCCACAGAATACT[A/C]GGAGTATTCAGAGTACTCATCATCC
Phenotype
MIM: 120120 MIM: 605320 MIM: 605902
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COL7A1 PubMed Links

Gene Details

Gene
COL7A1
Gene Name
collagen type VII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000094.3 8570 Nonsense Mutation GAG,TAG E2857* NP_000085.1
Gene
PFKFB4
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4
There are no transcripts associated with this gene.

Gene
UCN2
Gene Name
urocortin 2
There are no transcripts associated with this gene.

View Full Product Details