Product Details

SNP ID
rs4078456
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:154155652 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGCACTTGTTGTTTTCTCCAAA[C/T]ATCCAAACAGAATATAAACTCAAAG
Phenotype
MIM: 191030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP210L PubMed Links

Gene Details

Gene
NUP210L
Gene Name
nucleoporin 210 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159484.1 2864 Intron NP_001152956.1
NM_207308.2 2864 Intron NP_997191.2
XM_011510122.1 2864 Intron XP_011508424.1
XM_011510123.1 2864 Intron XP_011508425.1
XM_011510124.1 2864 Intron XP_011508426.1
XM_017002788.1 2864 Intron XP_016858277.1
XM_017002789.1 2864 Intron XP_016858278.1
Gene
TPM3
Gene Name
tropomyosin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043351.1 2864 UTR 3 NP_001036816.1
NM_001043352.1 2864 UTR 3 NP_001036817.1
NM_001043353.1 2864 UTR 3 NP_001036818.1
NM_001278188.1 2864 UTR 3 NP_001265117.1
NM_001278189.1 2864 UTR 3 NP_001265118.1
NM_001278190.1 2864 UTR 3 NP_001265119.1
NM_001278191.1 2864 UTR 3 NP_001265120.1
NM_152263.3 2864 Intron NP_689476.2
NM_153649.3 2864 UTR 3 NP_705935.1

View Full Product Details