Product Details

SNP ID
hCV27540213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:161166878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGACCGTGGTCGTGCTGGGCGGA[A/G]GCATCAGCGGCTTGGCCGCCAGTTA
Phenotype
MIM: 604014 MIM: 600923 MIM: 610554 MIM: 604729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALT3 PubMed Links

Gene Details

Gene
B4GALT3
Gene Name
beta-1,4-galactosyltransferase 3
There are no transcripts associated with this gene.

Gene
PPOX
Gene Name
protoporphyrinogen oxidase
There are no transcripts associated with this gene.

Gene
UFC1
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
There are no transcripts associated with this gene.

Gene
USP21
Gene Name
ubiquitin specific peptidase 21
There are no transcripts associated with this gene.

View Full Product Details