Product Details
- SNP ID
-
hCV27541507
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:46853695 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGTTTTTCCGGAATTGCAGAGAT[G/T]GCAAGTGCACATTAGCCTGCCAACA
- Phenotype
-
MIM: 300757
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RP2
PubMed Links
Gene Details
- Gene
- RP2
- Gene Name
- retinitis pigmentosa 2 (X-linked recessive)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006915.2 |
511 |
Missense Mutation |
GGC,TGC |
G108C |
NP_008846.2 |
View Full Product Details