Product Details

SNP ID
rs36018672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24055644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCAAGTTGACGCCACTTTCACA[C/T]GAAGTTATCAGCAGACAAGCCACAA
Phenotype
MIM: 300161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2S3 PubMed Links

Gene Details

Gene
EIF2S3
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001415.3 120 Silent Mutation CAC,CAT H33H NP_001406.1

View Full Product Details