Product Details

SNP ID
rs77000186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57543741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGAGCCCCACTGGGGAGGGG[C/T]ACAACCTCAGCACAGGTGAGTGAGA
Phenotype
MIM: 613664 MIM: 603890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105565.2 852 Missense Mutation CAC,TAC H284Y NP_001099035.2
XM_005273819.3 852 Missense Mutation CAC,TAC H284Y XP_005273876.1
Gene
UBE2L6
Gene Name
ubiquitin conjugating enzyme E2 L6
There are no transcripts associated with this gene.

View Full Product Details