Product Details

SNP ID
rs58690669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118364929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGAATTGGAAAATCTTAACCCA[A/C]AGTATTTTAAAATGTATTGTATTTG
Phenotype
MIM: 603753
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBE4A PubMed Links

Gene Details

Gene
UBE4A
Gene Name
ubiquitination factor E4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204077.1 Intron NP_001191006.1
NM_004788.3 Intron NP_004779.2

View Full Product Details