Product Details

SNP ID
rs17138766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37411350 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGAGGAAGACCAAAGCAGCACT[A/C]GTTGCCAATTAGGGAATGGACCGTT
Phenotype
MIM: 200350 MIM: 602276
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACACA PubMed Links

Gene Details

Gene
ACACA
Gene Name
acetyl-CoA carboxylase alpha
There are no transcripts associated with this gene.

Gene
TADA2A
Gene Name
transcriptional adaptor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166105.2 146 UTR 5 NP_001159577.2
NM_001291918.1 146 UTR 5 NP_001278847.1
NM_001488.4 146 UTR 5 NP_001479.4
NM_133439.3 146 UTR 5 NP_597683.3
XM_006722044.3 146 UTR 5 XP_006722107.1
XM_011525153.2 146 UTR 5 XP_011523455.1
XM_011525154.2 146 UTR 5 XP_011523456.1
XM_011525155.2 146 UTR 5 XP_011523457.1
XM_017024981.1 146 UTR 5 XP_016880470.1
XM_017024982.1 146 UTR 5 XP_016880471.1
XM_017024983.1 146 UTR 5 XP_016880472.1
XM_017024984.1 146 UTR 5 XP_016880473.1
XM_017024985.1 146 UTR 5 XP_016880474.1
XM_017024986.1 146 UTR 5 XP_016880475.1

View Full Product Details