Product Details

SNP ID
rs138099460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:57630867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTTCTGACTCATCCGGCAGAT[C/T]GGAACACCTGCATTTCTGGATGTGT
Phenotype
MIM: 611760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH17 PubMed Links

Gene Details

Gene
PCDH17
Gene Name
protocadherin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040429.2 764 Intron NP_001035519.1
XM_005266357.2 764 UTR 5 XP_005266414.1
XM_005266358.2 764 UTR 5 XP_005266415.1
XM_017020547.1 764 UTR 5 XP_016876036.1

View Full Product Details