Product Details

SNP ID
rs75702304
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:163325751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAAGCACGTGGTTTCTCATCT[A/G]TCTATTCAACCAAAGTCCTCCTTCC
Phenotype
MIM: 611772 MIM: 603276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUF2 PubMed Links

Gene Details

Gene
NUF2
Gene Name
NUF2, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031423.3 Intron NP_113611.2
NM_145697.2 Intron NP_663735.2
XM_011510036.1 Intron XP_011508338.1
Gene
RGS5
Gene Name
regulator of G-protein signaling 5
There are no transcripts associated with this gene.

View Full Product Details