Product Details

SNP ID
rs10497275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165990220 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCAATAAATACTGTGCTTAGGTC[A/G]TTATTTGTTTGCTCAAACGTGCACC
Phenotype
MIM: 182389
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724058 PubMed Links

Gene Details

Gene
LOC102724058
Gene Name
uncharacterized LOC102724058
There are no transcripts associated with this gene.

Gene
SCN1A
Gene Name
sodium voltage-gated channel alpha subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165963.1 7260 UTR 3 NP_001159435.1
NM_001165964.1 7260 UTR 3 NP_001159436.1
NM_001202435.1 7260 UTR 3 NP_001189364.1
NM_006920.4 7260 UTR 3 NP_008851.3
XM_011511602.2 7260 UTR 3 XP_011509904.1
XM_011511604.2 7260 Intron XP_011509906.1
XM_011511605.2 7260 UTR 3 XP_011509907.1
XM_011511606.2 7260 UTR 3 XP_011509908.1
XM_017004644.1 7260 UTR 3 XP_016860133.1
XM_017004645.1 7260 Intron XP_016860134.1
XM_017004646.1 7260 Intron XP_016860135.1
XM_017004647.1 7260 Intron XP_016860136.1
XM_017004648.1 7260 Intron XP_016860137.1
XM_017004649.1 7260 Intron XP_016860138.1
XM_017004650.1 7260 UTR 3 XP_016860139.1
XM_017004651.1 7260 UTR 3 XP_016860140.1
XM_017004652.1 7260 UTR 3 XP_016860141.1
XM_017004653.1 7260 UTR 3 XP_016860142.1
XM_017004654.1 7260 UTR 3 XP_016860143.1

View Full Product Details