Product Details

SNP ID
rs1055193216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69356267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCCTAATAGACTTCACCATTT[C/T]CTAGGAGAAGGTTCTACAGATTACC
Phenotype
MIM: 602027
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TERF2 PubMed Links

Gene Details

Gene
TERF2
Gene Name
telomeric repeat binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005652.4 2277 UTR 3 NP_005643.2
Gene
TMED6
Gene Name
transmembrane p24 trafficking protein 6
There are no transcripts associated with this gene.

View Full Product Details