Product Details

SNP ID
rs151245586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55947847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTGCAGGTCTCGTGTTTCTCT[C/T]TTCCAATCGGTTGTCTTTATCGTGG
Phenotype
MIM: 609659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NLRP8 PubMed Links

Gene Details

Gene
NLRP8
Gene Name
NLR family pyrin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317000.1 16 UTR 5 NP_001303929.1
NM_176811.2 16 UTR 5 NP_789781.2

View Full Product Details