Product Details

SNP ID
rs72737548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76826729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGACAAGCCTCTTCAGCCTGAGA[A/C]AGAAGGTGAAAGGGCATCCCATCTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC74A PubMed Links

Gene Details

Gene
LRRC74A
Gene Name
leucine rich repeat containing 74A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194287.2 294 Intron NP_919263.2
XM_011536479.1 294 UTR 5 XP_011534781.1
XM_011536483.1 294 UTR 5 XP_011534785.1
XM_011536487.1 294 UTR 5 XP_011534789.1
XM_017021032.1 294 Intron XP_016876521.1
XM_017021033.1 294 UTR 5 XP_016876522.1
XM_017021034.1 294 Intron XP_016876523.1
XM_017021035.1 294 Missense Mutation AAA,ACA K11T XP_016876524.1
XM_017021036.1 294 UTR 5 XP_016876525.1
XM_017021037.1 294 Intron XP_016876526.1
XM_017021038.1 294 Intron XP_016876527.1
XM_017021039.1 294 Intron XP_016876528.1
XM_017021040.1 294 Intron XP_016876529.1
XM_017021041.1 294 Intron XP_016876530.1

View Full Product Details