Product Details

SNP ID
rs7690308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:78553808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCAACTTGCATTTCTAAATCAA[A/C]CCTGGAGATATGCCAGCATGTTTGA
Phenotype
MIM: 106490 MIM: 607830
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANXA3 PubMed Links

Gene Details

Gene
ANXA3
Gene Name
annexin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005139.2 Intron NP_005130.1
Gene
FRAS1
Gene Name
Fraser extracellular matrix complex subunit 1
There are no transcripts associated with this gene.

View Full Product Details