Product Details

SNP ID
rs6809205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48637726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCATTCAATAAAAAGCAAAGAA[A/C]AATTCTGTACTTTGGGCCCCTGACG
Phenotype
MIM: 604264 MIM: 610068
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CELSR3 PubMed Links

Gene Details

Gene
CELSR3
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001407.2 10699 UTR 3 NP_001398.2
Gene
MIR4793
Gene Name
microRNA 4793
There are no transcripts associated with this gene.

Gene
MIR6824
Gene Name
microRNA 6824
There are no transcripts associated with this gene.

Gene
SLC26A6
Gene Name
solute carrier family 26 member 6
There are no transcripts associated with this gene.

View Full Product Details