Product Details

SNP ID
rs1032883127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99898205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTCCTGTTCCTATGAACACCT[C/G]AGCTGGCACAAAAGCTGAGTAGTTT
Phenotype
MIM: 603296
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB14 PubMed Links

Gene Details

Gene
DNAJB14
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031723.3 3946 UTR 3 NP_001026893.1
NM_001278310.1 3946 UTR 3 NP_001265239.1
NM_001278311.1 3946 Intron NP_001265240.1
Gene
LAMTOR3
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
There are no transcripts associated with this gene.

View Full Product Details