Product Details

SNP ID
rs61660861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55723223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGACAGAGCCGAGGGGACAGA[C/T]ACTCTAGGGTGGCAGAAACAGCTGT
Phenotype
MIM: 616334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM100 PubMed Links

Gene Details

Gene
TMEM100
Gene Name
transmembrane protein 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099640.1 Intron NP_001093110.1
NM_018286.2 Intron NP_060756.2
XM_017024814.1 Intron XP_016880303.1
XM_017024815.1 Intron XP_016880304.1
XM_017024816.1 Intron XP_016880305.1

View Full Product Details