Product Details

SNP ID
hCV27837589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151452261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCGACCTCTTCCGCTCCCTAGG[C/G]CCTGGGACCTCCCGGTCCCTTGCCT
Phenotype
MIM: 616332 MIM: 614917
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARMT1 PubMed Links

Gene Details

Gene
ARMT1
Gene Name
acidic residue methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286562.1 4 UTR 5 NP_001273491.1
NM_024573.2 4 UTR 5 NP_078849.1
Gene
RMND1
Gene Name
required for meiotic nuclear division 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271937.1 4 Intron NP_001258866.1
NM_017909.3 4 Intron NP_060379.2

View Full Product Details