Product Details

SNP ID
rs73833910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:94452696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAAAAGGAAGGGCGTGTGGAC[A/G]GGGCAGAGCTTGGAAGGAAAATTCG
Phenotype
MIM: 605904
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDLIM5 PubMed Links

Gene Details

Gene
PDLIM5
Gene Name
PDZ and LIM domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011513.3 Intron NP_001011513.3
NM_001011515.2 Intron NP_001011515.1
NM_001011516.2 Intron NP_001011516.1
NM_001256425.1 Intron NP_001243354.1
NM_001256426.1 Intron NP_001243355.1
NM_001256427.1 Intron NP_001243356.1
NM_001256428.1 Intron NP_001243357.1
NM_001256429.1 Intron NP_001243358.1
NM_006457.4 Intron NP_006448.4
XM_005262693.4 Intron XP_005262750.1
XM_005262695.4 Intron XP_005262752.1
XM_005262696.4 Intron XP_005262753.1
XM_005262698.4 Intron XP_005262755.1
XM_006714066.3 Intron XP_006714129.1
XM_006714068.3 Intron XP_006714131.1
XM_006714069.3 Intron XP_006714132.1
XM_006714070.3 Intron XP_006714133.1
XM_011531543.2 Intron XP_011529845.1
XM_017007657.1 Intron XP_016863146.1
XM_017007658.1 Intron XP_016863147.1

View Full Product Details