Product Details

SNP ID
rs113606579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21612319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGTGCTCCGCGCGCTCTTGA[C/G]GTCCGGAGCCCCTGGAGTAGGCGCT
Phenotype
MIM: 601063
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SNRPD1 PubMed Links

Gene Details

Gene
SNRPD1
Gene Name
small nuclear ribonucleoprotein D1 polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291916.1 51 UTR 5 NP_001278845.1
NM_006938.3 51 UTR 5 NP_008869.1

View Full Product Details