Product Details

SNP ID
rs6603441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118727358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGCGACGTGGTGGAAGAAGTTTT[G/T]TTCGGTCAGAGACCGGACGCGGGAG
Phenotype
MIM: 300119
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL13RA1 PubMed Links

Gene Details

Gene
IL13RA1
Gene Name
interleukin 13 receptor subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001560.2 405 Intron NP_001551.1
XM_011531336.1 405 Missense Mutation TTG,TTT L130F XP_011529638.1
XM_011531337.1 405 Intron XP_011529639.1
XM_017029507.1 405 Intron XP_016884996.1

View Full Product Details