Product Details

SNP ID
hCV27840349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:206897784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTCGCCAATTTAACACCAAGAA[G/T]AATTGAGGCTGCTTGGGAGGAAGGC
Phenotype
MIM: 606015 MIM: 604136
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
There are no transcripts associated with this gene.

Gene
IL24
Gene Name
interleukin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185156.1 227 UTR 5 NP_001172085.1
NM_001185157.1 227 UTR 5 NP_001172086.1
NM_001185158.1 227 UTR 5 NP_001172087.1
NM_006850.3 227 UTR 5 NP_006841.1

View Full Product Details