Product Details

SNP ID
rs76231002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89690839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAAGTTGTGGTTACCAAGGCGA[C/T]GCAACGCCGCCCGGCCAGGTGAGCA
Phenotype
MIM: 603866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEX11A PubMed Links

Gene Details

Gene
PEX11A
Gene Name
peroxisomal biogenesis factor 11 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271572.1 29 Intron NP_001258501.1
NM_001271573.1 29 Intron NP_001258502.1
NM_003847.2 29 Intron NP_003838.1
Gene
WDR93
Gene Name
WD repeat domain 93
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284395.1 29 UTR 5 NP_001271324.1
NM_001284396.1 29 UTR 5 NP_001271325.1
NM_020212.1 29 UTR 5 NP_064597.1
XM_006720614.1 29 UTR 5 XP_006720677.1
XM_006720615.1 29 UTR 5 XP_006720678.1
XM_006720618.2 29 UTR 5 XP_006720681.1
XM_011521794.2 29 Intron XP_011520096.1
XM_011521795.1 29 UTR 5 XP_011520097.1
XM_011521796.2 29 UTR 5 XP_011520098.1
XM_011521798.2 29 Intron XP_011520100.1
XM_011521800.1 29 UTR 5 XP_011520102.1
XM_011521802.2 29 UTR 5 XP_011520104.1
XM_017022415.1 29 UTR 5 XP_016877904.1
XM_017022416.1 29 UTR 5 XP_016877905.1
XM_017022417.1 29 UTR 5 XP_016877906.1

View Full Product Details