Product Details

SNP ID
rs35618443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:38208939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGGAACCCCGGAAGGAGCCACT[A/G]AAGTTGAGCAGAGTGCAGGACTGGG
Phenotype
MIM: 182451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSTR1 PubMed Links

Gene Details

Gene
SSTR1
Gene Name
somatostatin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001049.2 Intron NP_001040.1

View Full Product Details