Product Details

SNP ID
rs73518685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:73058462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGGACAAGAGCTGTTTTAAAT[G/T]TGTTTATCACACCGCCCCTAGTTTT
Phenotype
MIM: 602903
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLF5 PubMed Links

Gene Details

Gene
KLF5
Gene Name
Kruppel like factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286818.1 Intron NP_001273747.1
NM_001730.4 Intron NP_001721.2

View Full Product Details