Product Details

SNP ID
rs71651696
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:154567539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTGGCCCTCAGTACTAAGCAGG[C/G]CCTGACCTACTTGTCTTACTCTTGC
Phenotype
MIM: 118507
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 Intron NP_000739.1
XM_017000180.1 Intron XP_016855669.1
Gene
UBE2Q1
Gene Name
ubiquitin conjugating enzyme E2 Q1
There are no transcripts associated with this gene.

View Full Product Details