Product Details

SNP ID
rs6104289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45686272 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTAACAGAGTTTTTCTTTTCCA[A/G]AAGGTACTCTTTATTTCTGCTTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WFDC10B PubMed Links

Gene Details

Gene
WFDC10B
Gene Name
WAP four-disulfide core domain 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172006.2 277 Intron NP_742003.1
NM_172131.2 277 Intron NP_742143.1
XM_017027824.1 277 Intron XP_016883313.1
XM_017027825.1 277 UTR 5 XP_016883314.1

View Full Product Details