Product Details

SNP ID
rs33984246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116789734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGACCAGCTGTAGCAGTGGCC[A/G]CCAGGCAGCCAGAAGTGACTAGAGC
Phenotype
MIM: 606368 MIM: 603901
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOA5 PubMed Links

Gene Details

Gene
APOA5
Gene Name
apolipoprotein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166598.1 1568 UTR 3 NP_001160070.1
NM_052968.4 1568 UTR 3 NP_443200.2
XM_017017169.1 1568 UTR 3 XP_016872658.1
Gene
ZPR1
Gene Name
ZPR1 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317086.1 1568 Intron NP_001304015.1
NM_003904.4 1568 Intron NP_003895.1

View Full Product Details